siRNA Details
Gene Symbol: LOC684871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115043.1 | NP_001108515.1 | 1 | 144 | ||
GenBank mRNA | BC158773.1 | - | 1 | 144 | 7130655 |
Gene Symbol: LOC100910163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751622.3 | XP_003751670.1 | 510 |
