siRNA Details
Gene Symbol: LOC102550394 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FQ228322.1 | - | 1 | 440 |
Gene Symbol: LOC103694491 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773326.1 | XP_008771548.1 | 1 | 811 |
