siRNA Details
Gene Symbol: Dhx34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271452.1 | NP_001258381.1 | 6 | 1653 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dhx34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271452.1 | NP_001258381.1 | 6 | 1653 |