siRNA Details
Gene Symbol: LOC360919 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751340.3 | XP_003751388.1 | 9 | 1313 | ||
XM_006250759.2 | XP_006250821.1 | 9 | 1311 | |||
XM_006250760.2 | XP_006250822.1 | 9 | 1316 | |||
XM_006250761.2 | XP_006250823.1 | 7 | 1091 |
