siRNA Details
Gene Symbol: Cycs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012839.2 | NP_036971.1 | 3 | 680 | ||
GenBank mRNA | BC081849.1 | - | 3 | 680 | 7192255 | |
BC089051.1 | - | 3 | 691 | 7305562 | ||
FQ211631.1 | - | 3 | 672 | |||
FQ212556.1 | - | 3 | 674 | |||
FQ212860.1 | - | 3 | 732 | |||
FQ213464.1 | - | 3 | 668 | |||
FQ214646.1 | - | 3 | 703 | |||
FQ214733.1 | - | 3 | 672 | |||
FQ214738.1 | - | 3 | 670 | |||
FQ214838.1 | - | 3 | 671 | |||
FQ214855.1 | - | 3 | 703 | |||
FQ214908.1 | - | 3 | 672 | |||
FQ215052.1 | - | 3 | 672 | |||
FQ215109.1 | - | 3 | 672 | |||
FQ215466.1 | - | 3 | 672 | |||
FQ215858.1 | - | 3 | 715 | |||
FQ216163.1 | - | 3 | 674 | |||
FQ216217.1 | - | 3 | 670 | |||
FQ216281.1 | - | 3 | 730 | |||
FQ216512.1 | - | 3 | 672 | |||
FQ216560.1 | - | 3 | 673 | |||
FQ216674.1 | - | 3 | 671 | |||
FQ216679.1 | - | 3 | 672 | |||
FQ217283.1 | - | 3 | 672 | |||
FQ217634.1 | - | 3 | 672 | |||
FQ218428.1 | - | 3 | 672 | |||
FQ218668.1 | - | 3 | 673 | |||
FQ220222.1 | - | 3 | 703 | |||
FQ220855.1 | - | 3 | 715 | |||
FQ223761.1 | - | 3 | 672 | |||
FQ224710.1 | - | 3 | 672 | |||
FQ224919.1 | - | 3 | 673 |
Gene Symbol: LOC100363502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751799.3 | XP_003751847.1 | 1 | 676 |
