siRNA Details
Gene Symbol: Kif26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170348.1 | NP_001163819.1 | 7, 8 | 1564 | ||
XM_006240635.2 | XP_006240697.1 | 7, 8 | 1570 | |||
XM_006240636.2 | XP_006240698.1 | 7, 8 | 1570 | |||
XM_008764942.1 | XP_008763164.1 | 7, 8 | 1570 |
