siRNA Details
Gene Symbol: Kif26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170348.1 | NP_001163819.1 | 11 | 2266 | ||
XM_006240635.2 | XP_006240697.1 | 11 | 2272 | |||
XM_006240636.2 | XP_006240698.1 | 11 | 2272 | |||
XM_008764942.1 | XP_008763164.1 | 11 | 2272 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
