siRNA Details
Gene Symbol: Zfp322a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135084.1 | NP_001128556.1 | 4 | 827 | ||
XM_006253974.1 | XP_006254036.1 | 5 | 1150 | |||
XM_006253975.1 | XP_006254037.1 | 4 | 865 | |||
XM_008771661.1 | XP_008769883.1 | 3 | 2859 | |||
XM_008771662.1 | XP_008769884.1 | 3 | 1806 | |||
XM_008771663.1 | XP_008769885.1 | 4 | 1894 | |||
XM_008771664.1 | XP_008769886.1 | 4 | 2303 | |||
XM_008771665.1 | XP_008769887.1 | 4 | 1908 | |||
XM_008771666.1 | XP_008769888.1 | 4 | 2203 | |||
XM_008771667.1 | XP_008769889.1 | 4 | 2115 | |||
XM_008771668.1 | XP_008769890.1 | 4 | 2118 | |||
XM_008771669.1 | XP_008769891.1 | 4 | 1723 | |||
XM_008771670.1 | XP_008769892.1 | 4 | 1918 | |||
GenBank mRNA | FQ209962.1 | - | 4 | 637 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
