siRNA Details
Gene Symbol: Rnf169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001061888.5 | XP_001061888.3 | 4 | 1199 | ||
XM_006229823.2 | XP_006229885.1 | 3 | 2198 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rnf169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001061888.5 | XP_001061888.3 | 4 | 1199 | ||
XM_006229823.2 | XP_006229885.1 | 3 | 2198 |