siRNA Details
Gene Symbol: RGD1566006 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249195.2 | XP_006249257.2 | 1 | 61 | ||
XM_008769155.1 | XP_008767377.1 | 1 | 61 | |||
XM_008769156.1 | XP_008767378.1 | 1 | 61 |
