siRNA Details
Gene Symbol: Msrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044285.2 | NP_001037750.2 | 3 | 350 | ||
GenBank mRNA | BC094309.1 | - | 3 | 340 | 7316479 | |
FQ220312.1 | - | 3 | 362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
