siRNA Details
Gene Symbol: Cnpy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134710.1 | NP_001128182.1 | 2 | 257 | ||
GenBank mRNA | BC166762.1 | - | 2 | 257 | 7931606 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cnpy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134710.1 | NP_001128182.1 | 2 | 257 | ||
GenBank mRNA | BC166762.1 | - | 2 | 257 | 7931606 |