siRNA Details
Gene Symbol: Wfdc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253678.1 | NP_001240607.1 | 1, 2 | 122 | ||
XM_008762543.1 | XP_008760765.1 | 2, 3 | 199 | |||
GenBank mRNA | GQ406386.1 | - | 1, 2 | 122 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
