siRNA Details
Gene Symbol: Wfdc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253677.1 | NP_001240606.1 | 2 | 154 | ||
GenBank mRNA | GQ847861.1 | - | 2 | 154 |

More Information
Other Species Targeted

