siRNA Details
Gene Symbol: Wfdc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253677.1 | NP_001240606.1 | 2 | 150 | ||
GenBank mRNA | GQ847861.1 | - | 2 | 150 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
