siRNA Details
Gene Symbol: Efcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163921.1 | NP_001157393.1 | 6, 7 | 1489 | ||
XM_006236855.2 | XP_006236917.1 | 6, 7 | 2093 |

More Information
Other Species Targeted

Gene Ontology Categories:
