siRNA Details
Gene Symbol: Polr3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109468.1 | NP_001102938.1 | 2 | 744 | ||
XM_006231729.2 | XP_006231791.1 | 1 | 86 | |||
GenBank mRNA | FQ209718.1 | - | 2 | 503 | ||
FQ210837.1 | - | 2 | 503 |
