siRNA Details
Gene Symbol: Zfp385a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109470.2 | NP_001102940.1 | 6 | 832 | ||
NM_001135088.1 | NP_001128560.1 | 6 | 984 | |||
NM_001135089.1 | NP_001128561.1 | 6 | 883 | |||
XM_006242445.2 | XP_006242507.1 | 5 | 1009 | |||
XM_006242446.2 | XP_006242508.1 | 6 | 1189 | |||
GenBank mRNA | BC166729.1 | - | 6 | 984 | 7934026 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_013866, AF118566, AK137748, BC017644.
Gene Ontology Categories:
