siRNA Details
Gene Symbol: Rbbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107912.2 | NP_001101382.1 | 12 | 1701 | ||
GenBank mRNA | BC168994.1 | - | 12 | 1701 | 8362559 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rbbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107912.2 | NP_001101382.1 | 12 | 1701 | ||
GenBank mRNA | BC168994.1 | - | 12 | 1701 | 8362559 |