siRNA Details
Gene Symbol: Clgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109472.1 | NP_001102942.1 | 5, 6 | 639 | ||
XM_006255298.2 | XP_006255360.1 | 5, 6 | 592 | |||
XM_006255299.2 | XP_006255361.1 | 5, 6 | 673 |

More Information
Other Species Targeted

Gene Ontology Categories:
