siRNA Details
Gene Symbol: Frg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251429.2 | XP_006251491.1 | 3 | 519 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Frg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251429.2 | XP_006251491.1 | 3 | 519 |