siRNA Details
Gene Symbol: Gtf3c4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109473.1 | NP_001102943.1 | 2 | 1393 | ||
XM_006233871.2 | XP_006233933.1 | 2 | 1380 | |||
XM_006233872.2 | XP_006233934.1 | 2 | 2170 | |||
XM_008761680.1 | XP_008759902.1 | 3 | 1118 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
