siRNA Details
Gene Symbol: Gtf3c4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109473.1 | NP_001102943.1 | 4, 5 | 2514 | ||
XM_006233871.2 | XP_006233933.1 | 3, 4 | 2370 | |||
XM_006233872.2 | XP_006233934.1 | 4, 5 | 3291 | |||
XM_008761680.1 | XP_008759902.1 | 5, 6 | 2239 |

More Information
Other Species Targeted

Gene Ontology Categories:
