siRNA Details
Gene Symbol: LOC498316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109085.1 | NP_001102555.1 | 2 | 268 | ||
XM_006250489.2 | XP_006250551.1 | 2 | 328 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
