siRNA Details
Gene Symbol: Ggct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108629.2 | NP_001102099.1 | 4 | 574 | ||
XM_008762899.1 | XP_008761121.1 | 3 | 492 | |||
GenBank mRNA | FM060344.1 | - | 4 | 512 |
