siRNA Details
Gene Symbol: LOC685707 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751240.3 | XP_003751288.2 | 2 | 1611 | ||
XM_006249898.2 | XP_006249960.1 | 2 | 866 | |||
XM_006249899.2 | XP_006249961.1 | 2 | 866 | |||
XM_006249900.2 | XP_006249962.1 | 2 | 866 | |||
XM_006249902.2 | XP_006249964.1 | 2 | 865 |
