siRNA Details
Gene Symbol: Phemx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728846.4 | XP_002728892.3 | 5 | 2803 | ||
XM_006230915.2 | XP_006230977.1 | 4 | 2728 | |||
XM_006230916.2 | XP_006230978.1 | 5 | 2803 | |||
GenBank mRNA | BC158837.1 | - | 3 | 268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
