siRNA Details
Gene Symbol: Prp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768296.1 | XP_008766518.1 | 2 | 8089 | ||
GenBank mRNA | M86514.1 | - | 4936 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
