siRNA Details
Gene Symbol: Xpnpep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130582.2 | NP_001124054.2 | 3 | 254 | ||
XM_006242121.2 | XP_006242183.1 | 2 | 71 | |||
GenBank mRNA | BC168759.1 | - | 3 | 254 | 6888763 |

More Information
Other Species Targeted

Gene Ontology Categories:
