siRNA Details
Gene Symbol: LOC102550466 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770593.1 | XP_008768815.1 | 5 | 1213 |
Gene Symbol: LOC102546376 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770586.1 | XP_008768808.1 | 5 | 1308 |
