siRNA Details
Gene Symbol: Myl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109484.1 | NP_001102954.1 | 4 | 337 | ||
XM_008765039.1 | XP_008763261.1 | 4 | 331 | |||
XM_008765040.1 | XP_008763262.1 | 4 | 331 | |||
GenBank mRNA | BC166586.1 | - | 4 | 302 | 7934414 | |
FQ217612.1 | - | 4 | 298 | |||
FQ217952.1 | - | 4 | 310 | |||
FQ221192.1 | - | 4 | 304 | |||
FQ221499.1 | - | 4 | 306 | |||
FQ221709.1 | - | 4 | 338 | |||
FQ221718.1 | - | 4 | 309 | |||
FQ221751.1 | - | 4 | 305 | |||
FQ221963.1 | - | 4 | 306 | |||
FQ222034.1 | - | 4 | 297 | |||
FQ222160.1 | - | 4 | 307 | |||
FQ222384.1 | - | 4 | 309 | |||
FQ222504.1 | - | 4 | 286 | |||
FQ228533.1 | - | 4 | 309 |
Gene Symbol: Myl6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109486.1 | NP_001102956.1 | 4 | 337 |
Gene Symbol: Myl6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100983.1 | NP_001094453.1 | 3 | 281 | ||
GenBank mRNA | S77858.1 | - | 3 | 281 |

More Information
Other Species Targeted

Gene Ontology Categories:
