siRNA Details
Gene Symbol: Myl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109484.1 | NP_001102954.1 | 5 | 484 | ||
XM_008765039.1 | XP_008763261.1 | 5 | 478 | |||
XM_008765040.1 | XP_008763262.1 | 5 | 478 | |||
GenBank mRNA | BC166586.1 | - | 5 | 449 | 7934414 | |
FQ217612.1 | - | 5 | 445 | |||
FQ217952.1 | - | 5 | 457 | |||
FQ221005.1 | - | 5 | 455 | |||
FQ221192.1 | - | 5 | 451 | |||
FQ221499.1 | - | 5 | 453 | |||
FQ221709.1 | - | 5 | 485 | |||
FQ221718.1 | - | 5 | 456 | |||
FQ221751.1 | - | 5 | 452 | |||
FQ221963.1 | - | 5 | 453 | |||
FQ222034.1 | - | 5 | 444 | |||
FQ222160.1 | - | 5 | 454 | |||
FQ222384.1 | - | 5 | 456 | |||
FQ222504.1 | - | 5 | 433 | |||
FQ223317.1 | - | 5 | 459 | |||
FQ228533.1 | - | 5 | 456 |
Gene Symbol: Myl6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109486.1 | NP_001102956.1 | 5 | 384 |
Gene Symbol: Myl6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100983.1 | NP_001094453.1 | 4 | 428 | ||
GenBank mRNA | S77858.1 | - | 4 | 428 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
