siRNA Details
Gene Symbol: LOC498759 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY387080.1 | - | 1 | 471 |
Gene Symbol: LOC102548656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008764653.1 | XP_008762875.1 | 1 | 15 |
Gene Symbol: LOC102555068 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761995.1 | XP_008760217.1 | 1 | 15 |
Gene Symbol: LOC102557160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770720.1 | XP_008768942.1 | 1 | 15 |
Gene Symbol: LOC103690779 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768821.1 | XP_008767043.1 | 4 | 1101 |
Gene Symbol: LOC103694332 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772587.1 | XP_008770809.1 | 1 | 15 |
