siRNA Details
Gene Symbol: Sprr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001065728.2 | XP_001065728.1 | 1 | 384 | ||
XM_008761260.1 | XP_008759482.1 | 3 | 231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
