siRNA Details
Gene Symbol: Gapt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109491.1 | NP_001102961.1 | 2 | 713 | ||
XM_006231916.2 | XP_006231978.1 | 2 | 789 | |||
XM_006231917.2 | XP_006231979.1 | 2 | 749 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
