siRNA Details
Gene Symbol: TBC1D5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014744.2 | NP_055559.1 | 17, 18 | 3257 | ||
XM_005265614.1 | XP_005265671.1 | 17, 18 | 1827 | |||
XM_017007557.1 | XP_016863046.1 | 16, 17 | 1721 | |||
XM_017007558.1 | XP_016863047.1 | 16, 17 | 1751 | |||
XM_017007559.1 | XP_016863048.1 | 18, 19 | 2101 | |||
XM_017007560.1 | XP_016863049.1 | 18, 19 | 1931 | |||
XM_017007561.1 | XP_016863050.1 | 16, 17 | 1762 | |||
XM_017007563.1 | XP_016863052.1 | 15, 16 | 1681 | |||
XM_017007564.1 | XP_016863053.1 | 16, 17 | 1757 | |||
XM_017007565.1 | XP_016863054.1 | 17, 18 | 2055 | |||
XM_017007566.1 | XP_016863055.1 | 17, 18 | 1844 | |||
XM_017007567.1 | XP_016863056.1 | 15, 16 | 1685 | |||
GenBank mRNA | AB449886.1 | - | 15, 16 | 1592 | ||
AK097990.1 | - | 4, 5 | 701 | |||
BC013145.1 | - | 16, 17 | 1769 | 4510019 | ||
D86965.1 | - | 17, 18 | 3256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
