siRNA Details
Gene Symbol: LOC100912529 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750010.1 | XP_003750058.1 | 1 | 388 |
Gene Symbol: LOC686209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750016.1 | XP_003750064.1 | 388 |
