siRNA Details
Gene Symbol: LOC685619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729250.4 | XP_002729296.1 | 2 | 1045 | ||
XM_006235633.2 | XP_006235695.1 | 2 | 1042 |
Gene Symbol: Tp53rk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108606.1 | NP_001102076.1 | 973 |

More Information
Other Species Targeted

Gene Ontology Categories:
