siRNA Details
Gene Symbol: Fcrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232673.2 | XP_006232735.1 | 4 | 1473 | ||
XM_006232674.2 | XP_006232736.1 | 3 | 1452 | |||
GenBank mRNA | FQ232638.1 | - | 4 | 404 |

More Information
Other Species Targeted

