siRNA Details
Gene Symbol: Elmod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109506.1 | NP_001102976.1 | 2 | 209 | ||
XM_006255302.2 | XP_006255364.1 | 2 | 456 | |||
XM_006255303.2 | XP_006255365.1 | 2 | 255 |
Gene Symbol: LOC102554034 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772180.1 | XP_008770402.1 | 1 | 180 |

More Information
Other Species Targeted

Gene Ontology Categories:
