siRNA Details
Gene Symbol: Lrrn4cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109579.1 | NP_001103049.1 | 2 | 767 | ||
XM_006231002.1 | XP_006231064.1 | 2 | 2549 | |||
XM_006231004.2 | XP_006231066.1 | 2 | 2499 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
