siRNA Details
Gene Symbol: Amtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044296.1 | NP_001037761.1 | 8 | 644 | ||
XM_008770057.1 | XP_008768279.1 | 7 | 2300 | |||
GenBank mRNA | AM231717.1 | - | 8 | 604 | ||
DQ198381.1 | - | 8 | 644 |
