siRNA Details
Gene Symbol: Smarcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106861.1 | NP_001100331.1 | 14 | 1426 | ||
XM_006243770.2 | XP_006243832.1 | 14 | 1440 | |||
XM_006243771.2 | XP_006243833.1 | 14 | 1440 | |||
XM_006243772.2 | XP_006243834.1 | 14 | 1440 |

More Information
Other Species Targeted

Gene Ontology Categories:
