siRNA Details
Gene Symbol: Smarcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106861.1 | NP_001100331.1 | 16 | 1581 | ||
XM_006243770.2 | XP_006243832.1 | 16 | 1595 | |||
XM_006243771.2 | XP_006243833.1 | 16 | 1595 | |||
XM_006243772.2 | XP_006243834.1 | 16 | 1595 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
