siRNA Details
Gene Symbol: Stk38l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083336.1 | NP_001076805.1 | 14 | 1344 | ||
XM_008763431.1 | XP_008761653.1 | 13 | 2241 | |||
GenBank mRNA | BC160812.1 | - | 12 | 1226 | 7460150 | |
EF444939.1 | - | 14 | 1344 |
