siRNA Details
Gene Symbol: Slc25a36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766577.1 | XP_008764799.1 | 6 | 937 | ||
XM_576451.6 | XP_576451.4 | 7 | 1014 |
Gene Symbol: Slc25a36l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751866.3 | XP_003751914.2 | 3 | 952 |
