siRNA Details
Gene Symbol: LOC100363016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729879.3 | XP_002729925.1 | 2 | 625 |
Gene Symbol: LOC100912114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773716.1 | XP_008771938.1 | 2 | 690 |
