siRNA Details
Gene Symbol: SCRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145513.1 | NP_001138985.1 | 5 | 839 | ||
NM_001145514.1 | NP_001138986.1 | 5 | 771 | |||
NM_001145515.1 | NP_001138987.1 | 4 | 671 | |||
NM_014766.4 | NP_055581.3 | 5 | 831 | |||
GenBank mRNA | AB071705.1 | - | 5 | 721 | ||
AK289514.1 | - | 4 | 866 | |||
AK294891.1 | - | 2 | 201 | |||
AK295172.1 | - | 5 | 771 | |||
AK295713.1 | - | 4 | 574 | |||
BC040492.1 | - | 5 | 783 | 5314711 | ||
D83777.2 | - | 5 | 776 |
