siRNA Details
Gene Symbol: SPOCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244950.1 | NP_001231879.1 | 5 | 831 | ||
NM_014767.2 | NP_055582.1 | 6 | 714 | |||
XM_011540404.2 | XP_011538706.1 | 4 | 1045 | |||
GenBank mRNA | AJ001453.1 | - | 5 | 672 | ||
AK292328.1 | - | 5 | 564 | |||
AY358921.1 | - | 5 | 584 | |||
BC023558.2 | - | 5 | 825 | 4137728 | ||
D87465.1 | - | 6 | 702 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
