siRNA Details
Gene Symbol: Olr1421 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000780.1 | NP_001000780.1 | 1 | 879 |
Gene Symbol: LOC103690286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767749.1 | XP_008765971.1 | 1 | 861 |
